accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

ClHNOFFF

CAS RN 18706-22-4

IUPAC Name: 8-chloro-2-(trifluoromethyl)-1,4-dihydroquinolin-4-one
Synonyms: 8-chloro-2-(trifluoromethyl)-1H-quinolin-4-one
Molecular Weight (g/mol): 247.6
Molecular Formula: C10H5ClF3NO
InChi Key: SBVZSZNYPRWWNN-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(=O)C2=CC=CC(Cl)=C2N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results