Learn More
7-(4-Chlorobutoxy)-3,4-dihydro-2(1H)-quinolinone, Mikromol™
Product Code. 17941866
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17941866 25 mg 25mg
17952116 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17941866 Supplier Mikromol Supplier No. MM0961.060025
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
7-(4-Chlorobutoxy)-3,4-dihydro-2(1H)-quinolinone
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 7-(4-Chlorobutoxy)-3,4-dihydro-2(1H)-quinolinone
CAS 120004-79-7
Analyte or Component Names 7-(4-Chlorobutoxy)-3,4-dihydro-2(1H)-quinolinone
Shipping Condition Room Temperature
Molecular Formula C13 H16 Cl N O2
Certifications/Compliance ISO 9001
Synonym 7-(4-Chlorobutoxy)-3,4-dihydro-2(1H)-quinolinone, 7-(4-Chlorobutoxy)-3,4-dihydrocarbostyril
InChI Formula InChI=1S/C13H16ClNO2/c14-7-1-2-8-17-11-5-3-10-4-6-13(16)15-12(10)9-11/h3,5,9H,1-2,4,6-8H2,(H,15,16)
SMILES ClCCCCOc1ccc2CCC(=O)Nc2c1
IUPAC Name 7-(4-chlorobutoxy)-3,4-dihydro-1H-quinolin-2-one
Molecular Weight (g/mol) 253.72
Quantity 25 mg
Formula Weight 253.087 g/mol