Learn More
6Alpha-Hydroxy Ethynyl Estradiol, TRC
Product Code. 30133748
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133748 50 mg 50mg
30135397 10 mg 10mg
18013310 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30133748 Supplier Toronto Research Chemicals Supplier No. TRCH94200550MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 27521-34-2
Molecular Formula C20H24O3
Molecular Weight (g/mol) 312.4
Synonym 19-Nor-17alpha-pregna-1,3,5(10)-trien-20-yne-3,6alpha,17-triol, Ethinylestradiol Imp. E (EP), 6alpha-Hydroxyethinylestradiol
IUPAC Name (6S,8R,9S,13S,14S,17R)-17-ethynyl-13-methyl-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthrene-3,6,17-triol
SMILES C[C@]12CC[C@H]3[C@@H](C[C@H](O)c4cc(O)ccc34)[C@@H]1CC[C@@]2(O)C#C
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C20H24O3/c1-3-20(23)9-7-17-15-11-18(22)16-10-12(21)4-5-13(16)14(15)6-8-19(17,20)2/h1,4-5,10,14-15,17-18,21-23H,6-9,11H2,2H3/t14-,15-,17+,18+,19+,20+/m1/s1
Formula Weight 312.1725
Chemical Name or Material 19-Nor-17alpha-pregna-1,3,5(10)-trien-20-yne-3,6alpha,17-triol (6alpha-Hydroxyethinylestradiol)