Learn More
6alpha,9alpha-Difluoro-11beta,17alpha-dihydroxy-16alpha-methyl-3-oxoandrosta-1,4-diene-17beta-carboxylic Acid, Mikromol™
Description
6alpha,9alpha-Difluoro-11beta,17alpha-dihydroxy-16alpha-methyl-3-oxoandrosta-1,4-diene-17beta-carboxylic Acid
Specifications
Specifications
| Chemical Name or Material | 6alpha,9alpha-Difluoro-11beta,17alpha-dihydroxy-16alpha-methyl-3-oxoandrosta-1,4-diene-17beta-carboxylic Acid |
| CAS | 28416-82-2 |
| Analyte or Component Names | 6alpha,9alpha-Difluoro-11beta,17alpha-dihydroxy-16alpha-methyl-3-oxoandrosta-1,4-diene-17beta-carboxylic Acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H26 F2 O5 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 6alpha,9alpha-Difluoro-11beta,17alpha-dihydroxy-16alpha-methyl-3-oxoandrosta-1,4-diene-17beta-carboxylic Acid, Fluticasone 17beta-Carboxylic Acid, CCI 11400, Hydroxy-acid (fluticasone free acid intermediate), USP Fluticasone Propionate Related Compound J, GSK882497A, GSK862160A, GR243799X (21-oxidised flumethasone), CCI2857 (16-ß-methyl flumethasone), GSK495752A + CCI847, GW868300X (6-ß-fluoroflumethasone), CCI11400 (Hydroxy acid), GR164061X (9,11 epoxy flumethasone), GSK525421A (4-fluoro-6-des flumethasone), GSK1322629A, GSK657491A (hydroxyacid-17-formate), GR60001X (flumethasone acetate), Thioacid free acid (GR46580X), Thioacid furoate (GW675713X), Thioacid difuroate (GW725996X), 9,11 - epoxy hydroxyacid, GR51145X, 6-beta-fluoro hydroxyacid, GR144122X, 6-des-fluoro hydroxyacid, CCI4330, unidentified impurity with RRT 1.23, disulphide dipropionate dithioketone (GR 247095X), dithio (GR 269949X), Fluticasone Propionate Related Compound J |
| InChI Formula | InChI=1S/C21H26F2O5/c1-10-6-12-13-8-15(22)14-7-11(24)4-5-18(14,2)20(13,23)16(25)9-19(12,3)21(10,28)17(26)27/h4-5,7,10,12-13,15-16,25,28H,6,8-9H2,1-3H3,(H,26,27)/t10-,12+,13+,15+,16+,18+,19+,20+,21+/m1/s1 |
| SMILES | C[C@@H]1C[C@H]2[C@@H]3C[C@H](F)C4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)O |
| IUPAC Name | (6S,8S,9R,10S,11S,13S,14S,16R,17R)-6,9-difluoro-11,17-dihydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthrene-17-carboxylic acid |
| Show More |