missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6020A ICS Interferents A: Mo, Ti @ 20; Al, K, Mg, P, S @ 1000; C @ 2000; Fe, Na @ 2500; Ca @ 3000; Cl @ 20000 μg/mL in 2% HNO3, 6.5% HCl, tr. HF, VHG™
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Description
6020A ICS Interferents A: Mo, Ti @ 20; Al, K, Mg, P, S @ 1000; C @ 2000; Fe, Na @ 2500; Ca @ 3000; Cl @ 20000 μg/mL in 2% HNO3, 6.5% HCl, tr. HF
Specifications
Specifications
| Chemical Name or Material | Molybdenum/Titanium/Aluminium/Magnesium/Phosphorus/Potassium/Sulfur/Carbon/Iron/Sodium/Calcium/Chlorine |
| CAS | 7439-98-7, 7440-32-6, 7429-90-5, 7439-95-4, 7723-14-0, 9-7-7440 |
| Methods | ICP-MS |
| Analyte or Component Names | Molybdenum/Titanium/Aluminium/Magnesium/Phosphorus/Potassium/Sulfur/Carbon/Iron/Sodium/Calcium/Chlorine |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Dilute HNO3/HCl |
| Quantity | 500 mL |
| Concentration or Composition (by Analyte or Components) | Molybdenum 20 μg/mL/Titanium 20 μg/mL/Aluminium 1000 μg/mL/Magnesium 1000 μg/mL/Phosphorus 1000 μg/mL/Potassium 1000 μg/mL/Sulfur 1000 μg/mL/Carbon 2000 μg/mL/Iron 2500 μg/mL/Sodium 2500 μg/mL/Calcium 3000 μg/mL/Chlorine 20000 μg/mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction