Learn More
6-Methylidene-3,20-dioxopregn-4-en-17-yl Acetate (6-Methylenehydroxyprogesterone Acetate), Mikromol™
Description
6-Methylidene-3,20-dioxopregn-4-en-17-yl Acetate (6-Methylenehydroxyprogesterone Acetate)
Specifications
Specifications
| Chemical Name or Material | 6-Methylidene-3,20-dioxopregn-4-en-17-yl Acetate (6-Methylenehydroxyprogesterone Acetate) |
| CAS | 32634-95-0 |
| Analyte or Component Names | 6-Methylidene-3,20-dioxopregn-4-en-17-yl Acetate (6-Methylenehydroxyprogesterone Acetate) |
| Shipping Condition | Room Temperature |
| Molecular Formula | C24 H32 O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Medroxyprogesterone Acetate Imp. E (EP), Megestrol Acetate USP Related Compound D, Megestrol Acetate USP RC D, 6-Methylidene-3,20-dioxopregn-4-en-17-yl Acetate, 6-Methylenehydroxyprogesterone Acetate, Pregn-4-ene-3,20-dione, 17-(acetyloxy)-6-methylene-, Pregn-4-ene-3,20-dione, 17-hydroxy-6-methylene-, acetate (7CI,8CI), Progesterone, 17-hydroxy-6-methylene-, acetate (6CI), 17-(Acetyloxy)-6-methylenepregn-4-ene-3,20-dione, 17-Acetoxy-6-methylenepregn-4-ene-3,20-dione, 17alpha-Acetoxy-6-methylenepregn-4-ene-3,20-dione, 17alpha-Acetoxy-6-methyleneprogesterone, 6,6'-Dehydromedroxyprogesterone acetate, NSC 58799, Megestrol Acetate Related Compound D, Medroxyprogesterone Acetate Impurity E, Megestrol Acetate Impurity D |
| InChI Formula | InChI=1S/C24H32O4/c1-14-12-18-19(22(4)9-6-17(27)13-21(14)22)7-10-23(5)20(18)8-11-24(23,15(2)25)28-16(3)26/h13,18-20H,1,6-12H2,2-5H3/t18-,19+,20+,22-,23+,24+/m1/s1 |
| SMILES | CC(=O)O[C@@]1(CC[C@H]2[C@@H]3CC(=C)C4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C)C(=O)C |
| IUPAC Name | [(8R,9S,10R,13S,14S,17R)-17-acetyl-10,13-dimethyl-6-methylidene-3-oxo-1,2,7,8,9,11,12,14,15,16-decahydrocyclopenta[a]phenanthren-17-yl] acetate |
| Show More |