missing translation for 'onlineSavingsMsg'
Learn More

6-(Cyclopropylmethoxy)pyridine-3-boronic acid pinacol ester, 97%

Product Code. p-7033127
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11434610 250 mg 250mg
11444610 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11434610 Supplier Thermo Scientific Alfa Aesar Supplier No. H50139.MD
Add to basket
This item is not returnable. View return policy

CAS: 947191-69-7 | C15H22BNO3 | 275.155 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 947191-69-7
Molecular Formula C15H22BNO3
Molecular Weight (g/mol) 275.155
MDL Number MFCD07781184
InChI Key QEQNYJUPYICBCR-UHFFFAOYSA-N
Synonym 2-cyclopropylmethoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 6-cyclopropylmethoxy pyridine-3-boronic acid pinacol ester, 2-cyclopropylmethoxy pyridine-5-boronic acid, pinacol ester, 6-cyclopropylmethoxypyridine-3-boronic acid pinacol ester, 6-cyclopropylmethoxypyridine-3-yl boronic acid pinacol ester, 6-cyclopropylmethoxy pyridin-3-yl boronic acid pinacol ester, 2-cyclopropyl-methoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-cyclopropylmethoxy-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine, 2-cyclopropylmethoxy-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine
PubChem CID 24208797
IUPAC Name 2-(cyclopropylmethoxy)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CN=C(C=C2)OCC3CC3

Specifications

Quantity 250 mg
Sensitivity Air Sensitive
Formula Weight 275.16
Percent Purity 97%
Chemical Name or Material 6-(Cyclopropylmethoxy)pyridine-3-boronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.