accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

BOOCH3CH3H3CCH3NO

CAS RN 947191-69-7

IUPAC Name: 2-(cyclopropylmethoxy)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Synonyms: 2-(cyclopropylmethoxy)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Molecular Weight (g/mol): 275.16
Molecular Formula: C15H22BNO3
InChi Key: QEQNYJUPYICBCR-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CN=C(OCC2CC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results