Learn More
6-Chloropyrimidine-2,4-diamine 3-Oxide, Mikromol™
Product Code. 30052557
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30052557 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30052557 Supplier Mikromol Supplier No. MM0521.010025
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
6-Chloropyrimidine-2,4-diamine 3-Oxide
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 6-Chloropyrimidine-2,4-diamine 3-Oxide
CAS 35139-67-4
Analyte or Component Names 6-Chloropyrimidine-2,4-diamine 3-Oxide
Shipping Condition Room Temperature
Molecular Formula C4 H5 Cl N4 O
Certifications/Compliance ISO 9001
Synonym 6-Chloropyrimidine-2,4-diamine 3-Oxide, Minoxidil Imp. A (EP), Minoxidil Impurity A
InChI Formula InChI=1S/C4H5ClN4O/c5-2-1-3(6)9(10)4(7)8-2/h1H,6H2,(H2,7,8)
SMILES Nc1cc(Cl)nc(N)[n+]1[O-]
IUPAC Name 6-chloro-3-oxidopyrimidin-3-ium-2,4-diamine
Molecular Weight (g/mol) 160.56
Quantity 25 mg
Formula Weight 160.0152 g/mol