missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-2-cyclopropyl-4-(trifluoromethyl)-quinoline, TRC

Product Code. 30366922
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30366922 100 mg 100mg
30367892 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30366922 Supplier Toronto Research Chemicals Supplier No. TRCC365545100MG
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 391860-73-4
Molecular Formula C13 H9 Cl F3 N
Molecular Weight (g/mol) 271.67
Synonym 6-Chloro-2-cyclopropyl-4-(trifluoromethyl)quinoline, Efavirenz Related Compound C (USP)
IUPAC Name 6-chloro-2-cyclopropyl-4-(trifluoromethyl)quinoline
SMILES FC(F)(F)c1cc(nc2ccc(Cl)cc12)C3CC3

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C13H9ClF3N/c14-8-3-4-11-9(5-8)10(13(15,16)17)6-12(18-11)7-1-2-7/h3-7H,1-2H2
Formula Weight 271.0376
Chemical Name or Material 6-chloro-2-cyclopropyl-4-(trifluoromethyl)quinoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.