missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-4-hydroxyquinoline-3-carboxylic acid, 96%, Thermo Scientific™

Product Code. 15490738
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15490738

Brand: Thermo Scientific Alfa Aesar H54441.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents. Store at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98948-95-9
Molecular Formula C10H6BrNO3
Molecular Weight (g/mol) 268.066
MDL Number MFCD01569278
InChI Key GIUZUAUCCUFVKW-UHFFFAOYSA-N
Synonym 6-bromo-4-hydroxyquinoline-3-carboxylic acid, 6-bromo-4-oxo-1,4-dihydroquinoline-3-carboxylic acid, 6-bromo-4-hydroxy-3-quinolinecarboxylic acid, 4-hydroxy-6-bromoquinoline-3-carboxylic acid, 3-quinolinecarboxylic acid, 6-bromo-4-hydroxy, 3-quinolinecarboxylic acid, 6-bromo-1,4-dihydro-4-oxo, 6-bromo-4-hydroxyquinoline-3-carboxylicacid, 6-bromo-4-hydroxy-quinoline-3-carboxylic acid, 6-bromo-1,4-dihydro-4-oxoquinoline-3-carboxylic acid, pubchem17804
PubChem CID 693203
IUPAC Name 6-bromo-4-oxo-1H-quinoline-3-carboxylic acid
SMILES C1=CC2=C(C=C1Br)C(=O)C(=CN2)C(=O)O

Specifications

Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 268.06
Percent Purity 96%
Chemical Name or Material 6-Bromo-4-hydroxyquinoline-3-carboxylic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.