accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 98948-95-9

OHOHNBrO

CAS RN 98948-95-9

IUPAC Name: 6-bromo-4-oxo-1,4-dihydroquinoline-3-carboxylic acid
Synonyms: 6-bromo-4-oxo-1H-quinoline-3-carboxylic acid
Molecular Weight (g/mol): 268.07
Molecular Formula: C10H6BrNO3
InChi Key: GIUZUAUCCUFVKW-UHFFFAOYSA-N
SMILES: OC(=O)C1=CNC2=CC=C(Br)C=C2C1=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results