missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-2'-chloro-2,3'-bipyridine, 95%, Thermo Scientific™

Product Code. p-7029659
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11436270 100 mg 100mg
11446270 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11436270 Supplier Thermo Scientific Alfa Aesar Supplier No. H27149.MC

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 942206-02-2
Molecular Formula C10H6BrClN2
Molecular Weight (g/mol) 269.526
MDL Number MFCD09037439
InChI Key WFYCCIYVIHUIKA-UHFFFAOYSA-N
Synonym 6-bromo-2'-chloro-2,3'-bipyridine, 6-bromo-2'-chloro-2,3' bipyridine, 2-bromo-6-2-chloropyridin-3-yl pyridine
PubChem CID 42553003
IUPAC Name 2-bromo-6-(2-chloropyridin-3-yl)pyridine
SMILES C1=CC(=NC(=C1)Br)C2=C(N=CC=C2)Cl

Specifications

Melting Point 139°C to 142°C
Quantity 100 mg
Formula Weight 269.53
Percent Purity 95%
Chemical Name or Material 6-Bromo-2'-chloro-2,3'-bipyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.