accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

NBrNCl

CAS RN 942206-02-2

IUPAC Name: 6-bromo-2'-chloro-2,3'-bipyridine
Synonyms: 6-bromo-2'-chloro-2,3'-bipyridine
Molecular Weight (g/mol): 269.53
Molecular Formula: C10H6BrClN2
InChi Key: WFYCCIYVIHUIKA-UHFFFAOYSA-N
SMILES: ClC1=C(C=CC=N1)C1=NC(Br)=CC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results