Learn More
6-Beta-Hydroxytestosterone, Lipomed
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
6-Beta-Hydroxytestosterone
Specifications
Specifications
| Chemical Name or Material | 6 beta-Hydroxytestosterone |
| CAS | 62-99-7 |
| Analyte or Component Names | 6 beta-Hydroxytestosterone |
| Shipping Condition | Room Temperature |
| Molecular Formula | C19 H28 O3 |
| Synonym | Androst-4-en-3-one, 6,17-dihydroxy-, (6β,17β)-, Androst-4-en-3-one, 6β,17β-dihydroxy- (7 CI,8 CI), (6β,17β)-6,17-Dihydroxyandrost-4-en-3-one, 4-Androstene-6β,17β-diol-3-one, 6β,17β-Dihydroxy-4-androstene-3-one, 6β,17β-Dihydroxyandrost-4-en-3-one, 6β-Hydroxytestosterone, NSC 9721 |
| InChI Formula | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMILES | C[C@]12 CC[C@H]3 [C@@H](C[C@@H](O)C4=CC(=O)CC[C@]34 C)[C@@H]1 CC[C@@H]2 O |
| IUPAC Name | (6 R,8 R,9 S,10 R,13 S,14 S,17 S)-6,17-dihydroxy-10,13-dimethyl-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-3-one |
| Industry Type | Pharmaceutical |
| Show More |