missing translation for 'onlineSavingsMsg'
Learn More

N-Nitroso-L-proline, TRC

Product Code. 30005175
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005175 50 mg 50mg
17909634 5 mg 5mg
17997724 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30005175 Supplier Toronto Research Chemicals Supplier No. TRCN54545050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7519-36-0
Molecular Formula C5 H8 N2 O3
Molecular Weight (g/mol) 144.13
Synonym L-Proline, 1-nitroso- (6CI, 7CI, 9CI, ACI), 1-Nitroso-L-proline (ACI), Proline, 1-nitroso-, L- (8CI), N-Nitroso-L-proline, N-Nitrosoproline, NSC 109545, (2S)-1-Nitrosopyrrolidine-2-carboxylic acid
IUPAC Name (2S)-1-nitrosopyrrolidine-2-carboxylic acid
SMILES OC(=O)[C@@H]1CCCN1N=O

Specifications

InChI Formula InChI=1S/C5H8N2O3/c8-5(9)4-2-1-3-7(4)6-10/h4H,1-3H2,(H,8,9)/t4-/m0/s1
Quantity 50 mg
Formula Weight 144.0535
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material N-Nitroso-L-proline
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.