CAS RN 7519-36-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-Nitroso-L-proline, TRC
CAS: 7519-36-0 Molecular Formula: C5 H8 N2 O3 Molecular Weight (g/mol): 144.13 Synonym: L-Proline, 1-nitroso- (6CI, 7CI, 9CI, ACI),1-Nitroso-L-proline (ACI),Proline, 1-nitroso-, L- (8CI),N-Nitroso-L-proline,N-Nitrosoproline,NSC 109545,(2S)-1-Nitrosopyrrolidine-2-carboxylic acid IUPAC Name: (2S)-1-nitrosopyrrolidine-2-carboxylic acid SMILES: OC(=O)[C@@H]1CCCN1N=O