missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-[(RS)-[4-(Acetyloxy)phenyl]-(pyridin-2-yl)methyl]phenyl Acetate, Mikromol™
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
2-[(RS)-[4-(Acetyloxy)phenyl]-(pyridin-2-yl)methyl]phenyl Acetate
Specifications
Specifications
| Chemical Name or Material | 2-[(RS)-[4-(Acetyloxy)phenyl]-(pyridin-2-yl)methyl]phenyl Acetate |
| CAS | 111664-35-8 |
| Analyte or Component Names | 2-[(RS)-[4-(Acetyloxy)phenyl]-(pyridin-2-yl)methyl]phenyl Acetate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C22 H19 N O4 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Bisacodyl Imp. E (EP), Bisacodyl USP Related Compound E, Bisacodyl USP RC E, 2-[(RS)-[4-(Acetyloxy)phenyl]-(pyridin-2-yl)methyl]phenyl Acetate, Bisacodyl Related Compound E, Bisacodyl Impurity E |
| InChI Formula | InChI=1S/C22H19NO4/c1-15(24)26-18-12-10-17(11-13-18)22(20-8-5-6-14-23-20)19-7-3-4-9-21(19)27-16(2)25/h3-14,22H,1-2H3 |
| SMILES | CC(=O)Oc1ccc(cc1)C(c2ccccn2)c3ccccc3OC(=O)C |
| IUPAC Name | [4-[(2-acetyloxyphenyl)-pyridin-2-ylmethyl]phenyl] acetate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction