missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dihydroxybenzoic Acid-d3 (d2 Major), TRC

Product Code. 17917179
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17917179 50 mg 50mg
17977989 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 17917179 Supplier Toronto Research Chemicals Supplier No. TRCD45165750MG
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 294661-02-2
Molecular Formula C7H3D3O4
Molecular Weight (g/mol) 157.14
Synonym 5-Hydroxysalicylic Acid-d3, Carboxyhydroquinone-d3, Gensigen-d3, Gensigon-d3, Gentisinic Acid-d3, Hydroquinonecarboxylic Acid-d3, NSC 27224-d3, NSC 49098-d3, NSC 78825-d3;
SMILES OC1=C(C(O)=O)C([2H])=C(O)C([2H])=C1[2H]

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C7H6O4/c8-4-1-2-6(9)5(3-4)7(10)11/h1-3,8-9H,(H,10,11)/i1D,2D,3D
Chemical Name or Material 2,5-Dihydroxybenzoic Acid-d3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.