CAS RN 294661-02-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2,5-Dihydroxybenzoic Acid-d3 (d2 Major), TRC
CAS: 294661-02-2 Molecular Formula: C7H3D3O4 Molecular Weight (g/mol): 157.14 Synonym: 5-Hydroxysalicylic Acid-d3,Carboxyhydroquinone-d3,Gensigen-d3,Gensigon-d3,Gentisinic Acid-d3,Hydroquinonecarboxylic Acid-d3,NSC 27224-d3,NSC 49098-d3,NSC 78825-d3; SMILES: OC1=C(C(O)=O)C([2H])=C(O)C([2H])=C1[2H]