missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzylpenicillin Benzathine, Mikromol™
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Description
Benzylpenicillin Benzathine
Specifications
Specifications
| Chemical Name or Material | Benzathine benzylpenicillin |
| CAS | 1538-09-6 |
| Analyte or Component Names | Benzathine benzylpenicillin |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C16 H20 N2 . 2 C16 H18 N2 O4 S |
| Certifications/Compliance | ISO 17034 |
| Synonym | 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 3,3-dimethyl-7-oxo-6-[(2-phenylacetyl)amino]- (2S,5R,6R)-, compd. with N1,N2-bis(phenylmethyl)-1,2-ethanediamine (2:1), 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 3,3-dimethyl-7-oxo-6-(2-phenylacetamido)-, compd. with N,N'-dibenzylethylenediamine (2:1) (8CI), 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 3,3-dimethyl-7-oxo-6-[(phenylacetyl)amino]- (2S,5R,6R)-, compd. with N,N'-bis(phenylmethyl)-1,2-ethanediamine (2:1) (9CI), Penicillin G, N,N'-dibenzylethylenediamine salt (6CI), 1,2-Ethanediamine, N,N'-bis(phenylmethyl)-, bis[(2S,5R,6R)-3,3-dimethyl-7-oxo-6-[(phenylacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate] (9CI), Ethylenediamine, N,N'-dibenzyl-, bis[3,3-dimethyl-7-oxo-6-(2-phenylacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate] (8CI), Beacillin, Benepen, Benzacillin, Benzapen, Benzathine benzylpenicillin, Benzathine penicillin, Benzathine penicillin G, Benzethacil, Benzylpenicillin N,N'-dibenzylethylenediamine salt, Benzylpenicillin benzathine, Benzylpenicillin dibenzylethylenediamine salt, Bica-penicillin, Bicillin, Bicillin 1, Bicillin G, Bicillin I, Bicillin L-A, Cepacilina, Cillenta, Debecillin, Debecylina, Diamine penicillin, Dibencil, Dibencillin, Dibenzylethylenediamine dipenicillin G, Duropenin, Ethylenediamine, N,N'-dibenzyl-, compd. with penicillin G (1:2), Extencilline, Extenicilline, Lentopenil, Longacilina, Longicil, Megacillin suspension, Moldamin, N,N'-Dibenzylethylenediamine bis[benzyl penicillin], N,N'-Dibenzylethylenediamine salt of benzylpenicillin, Neolin, Pen-Di-Ben, Penadur, Pendepon, Penditan, Penduran, Penicillin G, compd. with N,N'-dibenzylethylenediamine (2:1), Penicom 12, Penidural, Penidure, Permapen, Strepdipen-Suspension, Tardocillin, Vicin, 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 3,3-dimethyl-7-oxo-6-[(phenylacetyl)amino]- [2S-(2alpha,5alpha,6beta)]-, compd. with N,N'-bis(phenylmethyl)-1,2-ethanediamine (2:1), 1,2-Ethanediamine, N,N'-bis(phenylmethyl)-, bis[[2S-(2alpha,5alpha,6beta)]-3,3-dimethyl-7-oxo-6-[(phenylacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate] |
| InChI Formula | InChI=1S/2C16H18N2O4S.C16H20N2/c2*1-16(2)12(15(21)22)18-13(20)11(14(18)23-16)17-10(19)8-9-6-4-3-5-7-9;1-3-7-15(8-4-1)13-17-11-12-18-14-16-9-5-2-6-10-16/h2*3-7,11-12,14H,8H2,1-2H3,(H,17,19)(H,21,22);1-10,17-18H,11-14H2/t2*11-,12+,14-;/m11./s1 |
| SMILES | CC1(C)S[C@@H]2[C@H](NC(=O)Cc3ccccc3)C(=O)N2[C@H]1C(=O)O.CC4(C)S[C@@H]5[C@H](NC(=O)Cc6ccccc6)C(=O)N5[C@H]4C(=O)O.C(CNCc7ccccc7)NCc8ccccc8 |
| IUPAC Name | N,N'-dibenzylethane-1,2-diamine;(2S,5R,6R)-3,3-dimethyl-7-oxo-6-[(2-phenylacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction