Learn More
5-Ketotilidine (Ethyl (1RS,2SR)-2-(Dimethylamino)-5-oxo-1-phenylcyclohex-3-enecarboxylate), Mikromol™
Product Code. 17993766
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17993766 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17993766 Supplier Mikromol Supplier No. MM0134.06
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
5-Ketotilidine (Ethyl (1RS,2SR)-2-(Dimethylamino)-5-oxo-1-phenylcyclohex-3-enecarboxylate)
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 5-Keto-Tilidine
CAS 2732673-20-8
Analyte or Component Names 5-Keto-Tilidine
Shipping Condition Ice pack (-20°C)
Molecular Formula C17 H21 N O3
Certifications/Compliance ISO 9001
Synonym 5-Ketotilidine, Ethyl (1RS,2SR)-2-(Dimethylamino)-5-oxo-1-phenylcyclohex-3-enecarboxylate, Ethyl (1R,6S)-6-(dimethylamino)-3-oxo-3,6-dihydro-[1,1'-biphenyl]-1(2H)-carboxylate, Ethyl rel-(1R,6S)-6-(dimethylamino)-3-oxo-3,6-dihydro-[1,1'-biphenyl]-1(2H)-carboxylate
InChI Formula InChI=1S/C17H21NO3/c1-4-21-16(20)17(13-8-6-5-7-9-13)12-14(19)10-11-15(17)18(2)3/h5-11,15H,4,12H2,1-3H3/t15-,17+/m0/s1
SMILES C(OCC)(=O)[C@@]1([C@@H](N(C)C)C=CC(=O)C1)C2=CC=CC=C2
IUPAC Name ethyl (1R,2S)-2-(dimethylamino)-5-oxo-1-phenylcyclohex-3-ene-1-carboxylate
Molecular Weight (g/mol) 287.35
Quantity 10 mg
Formula Weight 287.1521 g/mol
Mostrar más Mostrar menos
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.