Learn More
5-Hydroxytiabendazole, Mikromol™
Description
5-Hydroxytiabendazole
Specifications
Specifications
| Chemical Name or Material | Thiabendazole-5-hydroxy |
| CAS | 948-71-0 |
| Analyte or Component Names | Thiabendazole-5-hydroxy |
| Shipping Condition | Room Temperature |
| Molecular Formula | C10 H7 N3 O S |
| Certifications/Compliance | ISO 9001 |
| Synonym | 1H-Benzimidazol-5-ol, 2-(4-thiazolyl)- (9CI), 5(or 6)-Benzimidazolol, 2-(4-thiazolyl)- (7CI), 5-Benzimidazolol, 2-(4-thiazolyl)- (8CI), 2-(4-Thiazolyl)-1H-benzimidazol-6-ol, 5-Hydroxythiabendazole |
| InChI Formula | InChI=1S/C10H7N3OS/c14-6-1-2-7-8(3-6)13-10(12-7)9-4-15-5-11-9/h1-5,14H,(H,12,13) |
| SMILES | Oc1ccc2[nH]c(nc2c1)c3cscn3 |
| IUPAC Name | 2-(1,3-thiazol-4-yl)-1H-benzimidazol-5-ol |
| Show More |