Learn More
(5-(Difluoromethoxy)-1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl)methanesulfonic Acid Sodium Salt, TRC
Product Code. 30137682
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137682 100 mg 100mg
30011245 50 mg 50mg
30009824 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30137682 Supplier Toronto Research Chemicals Supplier No. TRCD367075100MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
Molecular Formula C7 H6 F5 N2 Na O4 S
Molecular Weight (g/mol) 332.18
IUPAC Name [5-(difluoromethoxy)-1-methyl-3-(trifluoromethyl)pyrazol-4-yl]methylsulfonyloxysodium
SMILES Cn1nc(c(CS(=O)(=O)O[Na])c1OC(F)F)C(F)(F)F
Specifications
Analyte or Component Names (5-(Difluoromethoxy)-1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl)methanesulfonic Acid Sodium Salt
Shipping Condition Room Temperature
InChI Formula InChI=1S/C7H7F5N2O4S.Na/c1-14-5(18-6(8)9)3(2-19(15,16)17)4(13-14)7(10,11)12;/h6H,2H2,1H3,(H,15,16,17);/q;+1/p-1
Solution Type Neat
Industry Type Pharmaceutical
Quantity 100 mg
Formula Weight 331.987
Chemical Name or Material (5-(Difluoromethoxy)-1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl)methanesulfonic Acid Sodium Salt