Learn More
5-Chlorohydrochlorothiazide, Mikromol™
Description
5-Chlorohydrochlorothiazide
Specifications
Specifications
| Chemical Name or Material | 5-Chloro Hydrochlorothiazide |
| CAS | 5233-42-1 |
| Analyte or Component Names | 5-Chloro Hydrochlorothiazide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C7 H7 Cl2 N3 O4 S2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 5-Chlorohydrochlorothiazide, 2H-1,2,4-Benzothiadiazine-7-sulfonamide, 5,6-dichloro-3,4-dihydro-, 1,1-dioxide, 5,6-Dichloro-3,4-dihydro-2H-1,2,4-benzothiadiazine-7-sulfonamide 1,1-dioxide |
| InChI Formula | InChI=1S/C7H7Cl2N3O4S2/c8-5-3(17(10,13)14)1-4-7(6(5)9)11-2-12-18(4,15)16/h1,11-12H,2H2,(H2,10,13,14) |
| SMILES | NS(=O)(=O)c1cc2c(NCNS2(=O)=O)c(Cl)c1Cl |
| IUPAC Name | 5,6-dichloro-1,1-dioxo-3,4-dihydro-2H-1λ^{6},2,4-benzothiadiazine-7-sulfonamide |
| Show More |