missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-methylphenyl isocyanate, 98%, Thermo Scientific™

Product Code. 11362497
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11362497

Brand: Thermo Scientific Alfa Aesar L11567.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40411-27-6
Molecular Formula C8H6ClNO
Molecular Weight (g/mol) 167.592
MDL Number MFCD00019913
InChI Key XEMUTFNBAICJEO-UHFFFAOYSA-N
Synonym 5-chloro-2-methylphenyl isocyanate, 5-chloro-2-methylphenylisocyanate, benzene,4-chloro-2-isocyanato-1-methyl, benzene, 4-chloro-2-isocyanato-1-methyl, 5-chloro-2-methylbenzenisocyanate, acmc-1aqw4, 4-chloro-2-isocyanatotoluene, 2-methyl-5-chlorophenylisocyanate, 5-chloro-1-isocyanato-2-methyl benzene, 4-chloranyl-2-isocyanato-1-methyl-benzene
PubChem CID 2769632
IUPAC Name 4-chloro-2-isocyanato-1-methylbenzene
SMILES CC1=C(C=C(C=C1)Cl)N=C=O

Specifications

Density 1.226
Boiling Point 115°C (5 mmHg)
Quantity 1 g
Refractive Index 1.556
UN Number UN2206
Beilstein 2691112
Formula Weight 167.6
Percent Purity 98%
Chemical Name or Material 5-Chloro-2-methylphenyl isocyanate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.