accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 40411-27-6

CH3ClNCO

CAS RN 40411-27-6

IUPAC Name: 4-chloro-2-isocyanato-1-methylbenzene
Synonyms: 4-chloro-2-isocyanato-1-methylbenzene
Molecular Weight (g/mol): 167.59
Molecular Formula: C8H6ClNO
InChi Key: XEMUTFNBAICJEO-UHFFFAOYSA-N
SMILES: CC1=C(C=C(Cl)C=C1)N=C=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results