missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-cyanopyridine, 95%, Thermo Scientific Chemicals

Product Code. p-7047680
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11375395

Brand: Thermo Scientific Alfa Aesar L19621.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

It is employed as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a pharmaceutical intermediate.

Solubility
Soluble in dichloromethane, ether, ethyl acetate and methanol

Notes
Store in cool place. Keep container tightly closed. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97483-77-7
Molecular Formula C6H3BrN2
Molecular Weight (g/mol) 183.008
MDL Number MFCD00234144
InChI Key DMSHUVBQFSNBBL-UHFFFAOYSA-N
Synonym 5-bromo-2-cyanopyridine, 5-bromopicolinonitrile, 5-bromo-2-pyridinecarbonitrile, 2-cyano-5-bromopyridine, 2-pyridinecarbonitrile, 5-bromo, 3-bromo-6-pyridinecarbonitrile, 5-bromo-pyridine-2-carbonitrile, 2-cyano-5-bromopyridin, pubchem2108, 5-bromo 2-cyanopyridine
PubChem CID 817154
IUPAC Name 5-bromopyridine-2-carbonitrile
SMILES C1=CC(=NC=C1Br)C#N

Specifications

Melting Point 124°C to 127°C
Quantity 1 g
Beilstein 5498624
Solubility Information Soluble in dichloromethane,ether,ethyl acetate and methanol
Formula Weight 183.01
Percent Purity 95%
Chemical Name or Material 5-Bromo-2-cyanopyridine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.