accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

BrNN

CAS RN 97483-77-7

IUPAC Name: 5-bromopyridine-2-carbonitrile
Synonyms: 5-bromopyridine-2-carbonitrile
Molecular Weight (g/mol): 183.01
Molecular Formula: C6H3BrN2
InChi Key: DMSHUVBQFSNBBL-UHFFFAOYSA-N
SMILES: BrC1=CC=C(N=C1)C#N
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)