Learn More
5-Bromo-2-cyanopyridine, 95%, Thermo Scientific Chemicals
Product Code. 11375395
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11375395 1 g 1g
11385395 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11375395 Supplier Thermo Scientific Alfa Aesar Supplier No. L19621.03
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
It is employed as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed as a pharmaceutical intermediate.
Solubility
Soluble in dichloromethane, ether, ethyl acetate and methanol
Notes
Store in cool place. Keep container tightly closed. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 97483-77-7
Molecular Formula C6H3BrN2
Molecular Weight (g/mol) 183.008
MDL Number MFCD00234144
InChI Key DMSHUVBQFSNBBL-UHFFFAOYSA-N
Synonym 5-bromo-2-cyanopyridine, 5-bromopicolinonitrile, 5-bromo-2-pyridinecarbonitrile, 2-cyano-5-bromopyridine, 2-pyridinecarbonitrile, 5-bromo, 3-bromo-6-pyridinecarbonitrile, 5-bromo-pyridine-2-carbonitrile, 2-cyano-5-bromopyridin, pubchem2108, 5-bromo 2-cyanopyridine
PubChem CID 817154
IUPAC Name 5-bromopyridine-2-carbonitrile
SMILES C1=CC(=NC=C1Br)C#N
Specifications
Melting Point 124°C to 127°C
Quantity 1 g
Beilstein 5498624
Solubility Information Soluble in dichloromethane,ether,ethyl acetate and methanol
Formula Weight 183.01
Percent Purity 95%
Chemical Name or Material 5-Bromo-2-cyanopyridine