Learn More
5-Bromo-2,4-difluorotoluene, 98%
Product Code. 15404847
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15404847 5 g 5g
15414847 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15404847 Supplier Thermo Scientific Alfa Aesar Supplier No. H32155.06
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
CAS: 159277-47-1 | C7H5BrF2 | 207.018 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 159277-47-1
Molecular Formula C7H5BrF2
Molecular Weight (g/mol) 207.018
MDL Number MFCD15144620
InChI Key WOCZKGUFVNUBEC-UHFFFAOYSA-N
Synonym 5-bromo-2,4-difluorotoluene, benzene, 1-bromo-2,4-difluoro-5-methyl, 1-methyl-2,4-difluoro-5-bromobenzene, 2,4-difluoro-5-bromo toluene
PubChem CID 10081748
IUPAC Name 1-bromo-2,4-difluoro-5-methylbenzene
SMILES CC1=CC(=C(C=C1F)F)Br
Specifications
Quantity 5 g
Formula Weight 207.02
Percent Purity 98%
Chemical Name or Material 5-Bromo-2,4-difluorotoluene