accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3FFBr

CAS RN 159277-47-1

IUPAC Name: 1-bromo-2,4-difluoro-5-methylbenzene
Synonyms: 1-bromo-2,4-difluoro-5-methylbenzene
Molecular Weight (g/mol): 207.02
Molecular Formula: C7H5BrF2
InChi Key: WOCZKGUFVNUBEC-UHFFFAOYSA-N
SMILES: CC1=CC(Br)=C(F)C=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results