missing translation for 'onlineSavingsMsg'
Learn More

5-Benzyl-1H-indazol-3-ol, TRC

Product Code. 30341200
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30341200 100 mg 100mg
30334489 25 mg 25mg
30331527 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30341200 Supplier Toronto Research Chemicals Supplier No. TRCB194830100MG
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Benzyl-1H-indazol-3-ol
Analyte or Component Names 5-Benzyl-1H-indazol-3-ol
Shipping Condition Room Temperature
Molecular Formula C14H12N2O
Synonym 5-Benzyl-1H-indazol-3-ol
InChI Formula InChI=1S/C14H12N2O/c17-14-12-9-11(6-7-13(12)15-16-14)8-10-4-2-1-3-5-10/h1-7,9H,8H2,(H2,15,16,17)
Solution Type Neat
SMILES OC1=NNC2=C1C=C(CC3=CC=CC=C3)C=C2
IUPAC Name 5-Benzyl-1H-indazol-3-ol
Industry Type Pharmaceutical
Molecular Weight (g/mol) 224.26
Quantity 100 mg
Formula Weight 224.09
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.