Learn More
5-Aminopyridine-2-carboxamidoxime, 97%, Thermo Scientific Chemicals
Product Code. 15470888
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15470888 5 g 5g
15480888 250 mg 250mg
15460888 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15470888 Supplier Thermo Scientific Alfa Aesar Supplier No. H54666.06
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1228776-03-1
Molecular Formula C6H8N4O
Molecular Weight (g/mol) 152.157
MDL Number MFCD20265317
InChI Key HDHDMYYDFKTNEQ-AATRIKPKSA-N
Synonym 2-pyridinecarboximidamide, 5-amino-n-hydroxy, 5-amino-n-hydroxypicolinimidamide
PubChem CID 58181821
IUPAC Name (6E)-6-[amino(nitroso)methylidene]-1H-pyridin-3-amine
SMILES C1=CC(=C(N)N=O)NC=C1N
Specifications
Quantity 5 g
Formula Weight 152.16
Percent Purity 97%
Chemical Name or Material 5-Aminopyridine-2-carboxamidoxime