CAS RN 1228776-03-1
CAS RN 1228776-03-1
IUPAC Name:
6-[amino(nitroso)methylidene]-1,6-dihydropyridin-3-amine
Synonyms:
6-[amino(nitroso)methylidene]-1H-pyridin-3-amine
Molecular Weight (g/mol):
152.16
Molecular Formula:
C6H8N4O
InChi Key:
HDHDMYYDFKTNEQ-UHFFFAOYSA-N
SMILES:
NC(N=O)=C1NC=C(N)C=C1
1
–
1
of
1
results
1
5-Aminopyridine-2-carboxamidoxime, 97%, Thermo Scientific Chemicals
CAS: 1228776-03-1 Molecular Formula: C6H8N4O Molecular Weight (g/mol): 152.157 MDL Number: MFCD20265317 InChI Key: HDHDMYYDFKTNEQ-AATRIKPKSA-N Synonym: 2-pyridinecarboximidamide, 5-amino-n-hydroxy,5-amino-n-hydroxypicolinimidamide PubChem CID: 58181821 IUPAC Name: (6E)-6-[amino(nitroso)methylidene]-1H-pyridin-3-amine SMILES: C1=CC(=C(N)N=O)NC=C1N