Learn More
5-Acetaminosalicylic Acid, Mikromol™
Product Code. 30049142
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30049142 25 mg 25mg
17961766 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30049142 Supplier Mikromol Supplier No. MM0217.020025
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
5-Acetaminosalicylic Acid
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-Acetyl Mesalazine
CAS 51-59-2
Analyte or Component Names N-Acetyl Mesalazine
Shipping Condition Room Temperature
Molecular Formula C9 H9 N O4
Certifications/Compliance ISO 17025
Synonym 5-Acetaminosalicylic Acid
InChI Formula InChI=1S/C9H9NO4/c1-5(11)10-6-2-3-8(12)7(4-6)9(13)14/h2-4,12H,1H3,(H,10,11)(H,13,14)
SMILES CC(=O)Nc1ccc(O)c(c1)C(=O)O
IUPAC Name 5-acetamido-2-hydroxybenzoic acid
Molecular Weight (g/mol) 195.17
Quantity 25 mg
Formula Weight 195.0532 g/mol