Learn More
5,6-Diamino-1,3-dimethyluracil, Mikromol™
Description
5,6-Diamino-1,3-dimethyluracil
Specifications
Specifications
| Chemical Name or Material | 5,6-Diamino-1,3-dimethyl Uracil |
| CAS | 5440-00-6 |
| Analyte or Component Names | 5,6-Diamino-1,3-dimethyl Uracil |
| Shipping Condition | Room Temperature |
| Molecular Formula | C6 H10 N4 O2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Uracil, 5,6-diamino-1,3-dimethyl- (6CI,7CI,8CI), 5,6-Diamino-1,3-dimethyl-2,4(1H,3H)-pyrimidinedione, 1,3-Dimethyl-5,6-diaminouracil, 4,5-Diamino-1,3-dimethyl-2,6-dioxotetrahydropyrimidine, 4,5-Diamino-1,3-dimethylpyrimidine-2,6-dione, 4,5-Diamino-1,3-dimethyluracil, 5,6-Diamino-1,3-dimethyl-2,4-pyrimidinedione, 5,6-Diamino-1,3-dimethyluracil, NSC 15493 |
| InChI Formula | InChI=1S/C6H10N4O2/c1-9-4(8)3(7)5(11)10(2)6(9)12/h7-8H2,1-2H3 |
| SMILES | CN1C(=C(N)C(=O)N(C)C1=O)N |
| IUPAC Name | 5,6-diamino-1,3-dimethylpyrimidine-2,4-dione |
| Show More |