Learn More
5-(2-Ethoxyphenyl)-1-methyl-3n-propyl-1,6-dihydro-7H-pyrazolo[4,3-d]pyrimidin-7-one, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
Description
5-(2-Ethoxyphenyl)-1-methyl-3n-propyl-1,6-dihydro-7H-pyrazolo[4,3-d]pyrimidin-7-one
Specifications
Specifications
| Chemical Name or Material | 5-(2-Ethoxyphenyl)-1-methyl-3n-propyl-1,6-dihydro-7H-pyrazolo[4,3-d]pyrimidin-7-one |
| CAS | 139756-21-1 |
| Analyte or Component Names | 5-(2-Ethoxyphenyl)-1-methyl-3n-propyl-1,6-dihydro-7H-pyrazolo[4,3-d]pyrimidin-7-one |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H20 N4 O2 |
| Certifications/Compliance | ISO 17025 |
| Synonym | 5-(2-Ethoxyphenyl)-1-methyl-3n-propyl-1,6-dihydro-7H-pyrazolo[4,3-d]pyrimidin-7-one |
| InChI Formula | InChI=1S/C17H20N4O2/c1-4-8-12-14-15(21(3)20-12)17(22)19-16(18-14)11-9-6-7-10-13(11)23-5-2/h6-7,9-10H,4-5,8H2,1-3H3,(H,18,19,22) |
| SMILES | CCCc1nn(C)c2C(=O)NC(=Nc12)c3ccccc3OCC |
| IUPAC Name | 5-(2-ethoxyphenyl)-1-methyl-3-propyl-6H-pyrazolo[4,3-d]pyrimidin-7-one |
| Show More |