Learn More
(4S)-2-[5-(4-Hydroxyphenyl)-3,6-dioxopiperazin-2-yl]-5,5-dimethylthiazolidine-4-carboxylic Acid (Amoxicillin Diketopiperazines), Mikromol™
Product Code. 17208509
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17208509 100 mg 100mg
30050275 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17208509 Supplier Mikromol Supplier No. MM0232.03
 Restricted Item
Estimated Shipment: 21-07-2026
Add to basket
Add to basket
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(4S)-2-[5-(4-Hydroxyphenyl)-3,6-dioxopiperazin-2-yl]-5,5-dimethylthiazolidine-4-carboxylic Acid (Amoxicillin Diketopiperazines)
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (4S)-2-[5-(4-Hydroxyphenyl)-3,6-dioxopiperazin-2-yl]-5,5-dimethylthiazolidine-4-carboxylic Acid (Amoxicillin Diketopiperazines)
CAS 94659-47-9
Analyte or Component Names (4S)-2-[5-(4-Hydroxyphenyl)-3,6-dioxopiperazin-2-yl]-5,5-dimethylthiazolidine-4-carboxylic Acid (Amoxicillin Diketopiperazines)
Shipping Condition Room Temperature
Molecular Formula C16 H19 N3 O5 S
Certifications/Compliance ISO 9001
Synonym Amoxicillin Trihydrate Imp. C (EP), Amoxicillin Imp. C (EP), Amoxicillin USP Related Compound C, Amoxicillin USP RC C, (4S)-2-[5-(4-Hydroxyphenyl)-3,6-dioxopiperazin-2-yl]-5,5-dimethylthiazolidine-4-carboxylic Acid, Amoxicillin Diketopiperazines, SB-206891, Ph Eur Amoxicillin Impurity C, Amoxicillin Related Compound C, Amoxicillin Trihydrate Impurity C, Amoxicillin Impurity C
InChI Formula InChI=1S/C16H19N3O5S/c1-16(2)11(15(23)24)19-14(25-16)10-13(22)17-9(12(21)18-10)7-3-5-8(20)6-4-7/h3-6,9-11,14,19-20H,1-2H3,(H,17,22)(H,18,21)(H,23,24)/t9?,10?,11-,14?/m0/s1
SMILES CC1(C)SC(N[C@H]1C(=O)O)C2NC(=O)C(NC2=O)c3ccc(O)cc3
IUPAC Name (4S)-2-[5-(4-hydroxyphenyl)-3,6-dioxopiperazin-2-yl]-5,5-dimethyl-1,3-thiazolidine-4-carboxylic acid
Molecular Weight (g/mol) 365.4
Quantity 100 mg
Formula Weight 365.1045 g/mol