Learn More
(4RS)-2-(1,1-Dimethylethyl)-1,2,3,4-tetrahydroisoquinoline-4,6,8-triol Hemisulphate, Mikromol™
Description
(4RS)-2-(1,1-Dimethylethyl)-1,2,3,4-tetrahydroisoquinoline-4,6,8-triol Hemisulphate
Specifications
Specifications
| Chemical Name or Material | (4RS)-2-(1,1-Dimethylethyl)-1,2,3,4-tetrahydroisoquinoline-4,6,8-triol Hemisulphate |
| CAS | 2732347-49-6 |
| Analyte or Component Names | (4RS)-2-(1,1-Dimethylethyl)-1,2,3,4-tetrahydroisoquinoline-4,6,8-triol Hemisulphate |
| Shipping Condition | Room Temperature |
| Molecular Formula | 2 C13 H19 N O3 . H2 O4 S |
| Certifications/Compliance | ISO 17025 |
| Synonym | Terbutaline Sulphate Imp. B (EP), Terbutaline Imp. B (EP), (4RS)-2-(1,1-Dimethylethyl)-1,2,3,4-tetrahydroisoquinoline-4,6,8-triol Hemisulphate, Terbutaline Sulphate Imp. B (EP) as Hemisulphate, Terbutaline Sulfate Impurity B as Hemisulfate, Terbutaline Impurity B as Hemisulfate |
| InChI Formula | InChI=1S/2C13H19NO3.H2O4S/c2*1-13(2,3)14-6-10-9(12(17)7-14)4-8(15)5-11(10)16;1-5(2,3)4/h2*4-5,12,15-17H,6-7H2,1-3H3;(H2,1,2,3,4) |
| SMILES | CC(C)(C)N1CC(O)c2cc(O)cc(O)c2C1.CC(C)(C)N3CC(O)c4cc(O)cc(O)c4C3.OS(=O)(=O)O |
| IUPAC Name | 2-tert-butyl-3,4-dihydro-1H-isoquinoline-4,6,8-triol;sulfuric acid |
| Show More |