Learn More
465 mV Redox Oxidation/Reduction (ORP) Standard at 25°C, Reagecon
Shop All Reagecon Products
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Description
- Redox Reference Standard, Redox Reference Material.
- Acts as a control or reference material rather than a calibration standard because the redox reaction is an absolute measurement and the sensing electrode does not require calibration.
- Controls the functionality of the sensing and reference electrode, the analysts technique, environmental conditions and the operation of the measuring meter.
- This 465 mV Redox (ORP) Standard is accurate to ± 5mV, has guaranteed stability throughout its entire shelf life, even after opening the bottle.
- The Value of 465 mV quoted refers to the potentials of the platinum electrode v Ag/AgCl reference electrode (3M KCl).
Specifications
Specifications
| Description | Accuracy ± 5mV at 25°C |
| Packaging | HDPE Bottle GL45 |
| Concentration or Composition Notes | 465 mV (± 5mV) |
| Physical Form | Liquid |
| Concentration | 465 mV (±5mV) |
| Quantity | 500 mL |
Safety and Handling
Product Identifier
- REDOX STANDARD 465mV a 25°C Ag/AgCl
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- H318-Causes serious eye damage.
- H315-Causes skin irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash thoroughly after handling.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P321-Specific treatment (see on this label).
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- MIXTURE LIST-Contains : sulphuric acid, irontrichloride, iron sulphate