missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)benzyl chloride, 98%, Thermo Scientific™

Product Code. 11459876
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11459876 1 g 1g
11469876 5 g 5g
11479876 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11459876 Supplier Thermo Scientific Alfa Aesar Supplier No. B20861.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 939-99-1
Molecular Formula C8H6ClF3
Molecular Weight (g/mol) 194.581
MDL Number MFCD00040772
InChI Key MCHDHQVROPEJJT-UHFFFAOYSA-N
Synonym 4-trifluoromethyl benzyl chloride, 4-trifluoromethylbenzyl chloride, 1-chloromethyl-4-trifluoromethyl benzene, p-trifluoromethylbenzyl chloride, ccris 5110, 4-chloromethyl benzotrifluoride, p-trifluoromethyl benzyl chloride, benzene, 1-chloromethyl-4-trifluoromethyl, alpha'-chloro-alpha,alpha,alpha-trifluoro-p-xylene
PubChem CID 123096
IUPAC Name 1-(chloromethyl)-4-(trifluoromethyl)benzene
SMILES C1=CC(=CC=C1CCl)C(F)(F)F

Specifications

Boiling Point 68°C (12 mmHg)
Flash Point 74°C (165°F)
Odor Pungent
Quantity 1 g
UN Number UN3265
Beilstein 511085
Formula Weight 194.59
Percent Purity 98%
Chemical Name or Material 4-(Trifluoromethyl)benzyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.