CAS RN 939-99-1
IUPAC Name:
1-(chloromethyl)-4-(trifluoromethyl)benzene
Synonyms:
1-(chloromethyl)-4-(trifluoromethyl)benzene
Molecular Weight (g/mol):
194.58
Molecular Formula:
C8H6ClF3
InChi Key:
MCHDHQVROPEJJT-UHFFFAOYSA-N
SMILES:
FC(F)(F)C1=CC=C(CCl)C=C1
1
–
1
of
1
results
1
4-(Trifluoromethyl)benzyl chloride, 98%, Thermo Scientific™
CAS: 939-99-1 Molecular Formula: C8H6ClF3 Molecular Weight (g/mol): 194.581 MDL Number: MFCD00040772 InChI Key: MCHDHQVROPEJJT-UHFFFAOYSA-N Synonym: 4-trifluoromethyl benzyl chloride,4-trifluoromethylbenzyl chloride,1-chloromethyl-4-trifluoromethyl benzene,p-trifluoromethylbenzyl chloride,ccris 5110,4-chloromethyl benzotrifluoride,p-trifluoromethyl benzyl chloride,benzene, 1-chloromethyl-4-trifluoromethyl,alpha'-chloro-alpha,alpha,alpha-trifluoro-p-xylene PubChem CID: 123096 IUPAC Name: 1-(chloromethyl)-4-(trifluoromethyl)benzene SMILES: C1=CC(=CC=C1CCl)C(F)(F)F