missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butyl-o-xylene, 98%, Thermo Scientific™

Product Code. 11382576
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11382576 25 g 25g
11372576 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11382576 Supplier Thermo Scientific Alfa Aesar Supplier No. L08552.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7397-06-0
Molecular Formula C12H18
Molecular Weight (g/mol) 162.276
MDL Number MFCD00038277
InChI Key QRPPSTNABSMSCS-UHFFFAOYSA-N
Synonym 4-tert-butyl-o-xylene, 4-tert-butyl-1,2-dimethylbenzene, 4-t-butyl-o-xylene, benzene, 4-1,1-dimethylethyl-1,2-dimethyl, acmc-209ot4, o-xylene, 4-tert-butyl, as-p2, 1,2-dimethyl-4-t-butylbenzene, 3,4-dimethyl-tert-butylbenzene
PubChem CID 81881
IUPAC Name 4-tert-butyl-1,2-dimethylbenzene
SMILES CC1=C(C=C(C=C1)C(C)(C)C)C

Specifications

Density 0.868
Boiling Point 211°C
Flash Point 82°C (179°F)
Refractive Index 1.499
Quantity 25 g
Beilstein 1925371
Formula Weight 162.27
Percent Purity 98%
Chemical Name or Material 4-tert-Butyl-o-xylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.