accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3H3CCH3H3CCH3

CAS RN 7397-06-0

IUPAC Name: 4-tert-butyl-1,2-dimethylbenzene
Synonyms: 4-tert-butyl-1,2-dimethylbenzene
Molecular Weight (g/mol): 162.28
Molecular Formula: C12H18
InChi Key: QRPPSTNABSMSCS-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1C)C(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results