Learn More
4-Nitrobenzo-18-crown-6, 99%
Product Code. 11423187
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 11423187
Brand: Thermo Scientific Alfa Aesar A16138.03
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
CAS: 53408-96-1 | C16H23NO8 | 357.359 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 53408-96-1
Molecular Formula C16H23NO8
Molecular Weight (g/mol) 357.359
MDL Number MFCD00143207
InChI Key LQXOKBZWNFJJGI-UHFFFAOYSA-N
Synonym 4-nitrobenzo-18-crown-6, 4'-nitrobenzo-18-crown 6-ether, nitrobenzo-18-crown-6, 18-nitro-2,3,5,6,8,9,11,12,14,15-decahydro-1,4,7,10,13,16-benzohexaoxacyclooctadecine, 20-nitro-2,5,8,11,14,17-hexaoxabicyclo 16.4.0 docosa-1 18 ,19,21-triene, 4'-nitrobenzo-18-crown-6, lqxokbzwnfjjgi-uhfffaoysa, 2,3-4-nitrobenzo-1,4,7,10,13,16-hexaoxacycloocta, 2,3-4-nitrobenzo-1,4,7,10,13,16-hexaoxacyclooctadec-2-ene
PubChem CID 602198
IUPAC Name 20-nitro-2,5,8,11,14,17-hexaoxabicyclo[16.4.0]docosa-1(18),19,21-triene
SMILES C1COCCOCCOC2=C(C=CC(=C2)[N+](=O)[O-])OCCOCCO1
Specifications
Melting Point 84°C to 86°C
Quantity 1 g
Formula Weight 357.35
Percent Purity 99%
Chemical Name or Material 4-Nitrobenzo-18-crown-6