accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 53408-96-1

ONOOOOOOO

CAS RN 53408-96-1

IUPAC Name: 18-nitro-2,3,5,6,8,9,11,12,14,15-decahydro-1,4,7,10,13,16-benzohexaoxacyclooctadecine
Synonyms: 18-nitro-2,3,5,6,8,9,11,12,14,15-decahydro-1,4,7,10,13,16-benzohexaoxacyclooctadecine
Molecular Weight (g/mol): 357.36
Molecular Formula: C16H23NO8
InChi Key: LQXOKBZWNFJJGI-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C2OCCOCCOCCOCCOCCOC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results