Learn More
4-n-Pentylaniline, 98%
Product Code. 11466010
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11466010 25 g 25g
11456010 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11466010 Supplier Thermo Scientific Alfa Aesar Supplier No. H26569.14
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
missing translation for 'validMainframeMsg'
CAS: 33228-44-3 | C11H17N | 163.264 g/mol
4-Pentylaniline is used in the synthesis of 4-[(4?-pentyl)azo]phenol (5Az-OH) by diazo-coupling reaction with phenol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Pentylaniline is used in the synthesis of 4-[(4′-pentyl)azo]phenol (5Az-OH) by diazo-coupling reaction with phenol
Solubility
Sparingly soluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 33228-44-3
Molecular Formula C11H17N
Molecular Weight (g/mol) 163.264
MDL Number MFCD00007926
InChI Key DGFTWBUZRHAHTH-UHFFFAOYSA-N
Synonym benzenamine, 4-pentyl, 4-amylaniline, p-pentylaniline, p-amylaniline, unii-8roj0g9hiu, p-n-pentylaniline, 4-n-pentylaniline, 4-pentylphenylamine, 8roj0g9hiu, p-n-amylaniline
PubChem CID 93162
IUPAC Name 4-pentylaniline
SMILES CCCCCC1=CC=C(C=C1)N
Specifications
Density 0.919
Boiling Point 130°C
Flash Point >110°C (230°F)
Refractive Index 1.53
Quantity 25 g
Solubility Information Sparingly soluble in water.
Formula Weight 163.26
Percent Purity 98%
Chemical Name or Material 4-n-Pentylaniline