accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 33228-44-3

IUPAC Name: 4-pentylaniline
Synonyms: benzenamine, 4-pentyl-
Molecular Weight (g/mol): 163.26
Molecular Formula: C11H17N
InChi Key: DGFTWBUZRHAHTH-UHFFFAOYSA-N
SMILES: CCCCCC1=CC=C(N)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results