Learn More
4-n-Pentylaniline, 98%
Product Code. 11456010
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11456010 5 g 5g
11466010 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11456010 Supplier Thermo Scientific Alfa Aesar Supplier No. H26569.06
 Restricted Item
In Stock
Add to basket
missing translation for 'validMainframeMsg'
CAS: 33228-44-3 | C11H17N | 163.264 g/mol
4-Pentylaniline is used in the synthesis of 4-[(4?-pentyl)azo]phenol (5Az-OH) by diazo-coupling reaction with phenol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Pentylaniline is used in the synthesis of 4-[(4′-pentyl)azo]phenol (5Az-OH) by diazo-coupling reaction with phenol
Solubility
Sparingly soluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 33228-44-3
Molecular Formula C11H17N
Molecular Weight (g/mol) 163.264
MDL Number MFCD00007926
InChI Key DGFTWBUZRHAHTH-UHFFFAOYSA-N
Synonym benzenamine, 4-pentyl, 4-amylaniline, p-pentylaniline, p-amylaniline, unii-8roj0g9hiu, p-n-pentylaniline, 4-n-pentylaniline, 4-pentylphenylamine, 8roj0g9hiu, p-n-amylaniline
PubChem CID 93162
IUPAC Name 4-pentylaniline
SMILES CCCCCC1=CC=C(C=C1)N
Specifications
Density 0.919
Boiling Point 130°C
Flash Point >110°C (230°F)
Refractive Index 1.53
Quantity 5 g
Solubility Information Sparingly soluble in water.
Formula Weight 163.26
Percent Purity 98%
Chemical Name or Material 4-n-Pentylaniline
compliance-icons
Product Identifier
  • 4-n-Pentylaniline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 4-pentyl-benzenamine